5-Methyl-thiophene-3-carboxylic acid

Supplier: Apollo Scientific
Warning

OR5951-1G OR5951-250MG OR5951-5G
APOSOR5951-1GEA 52.2 EUR
APOSOR5951-1G APOSOR5951-250MG APOSOR5951-5G
5-Methyl-thiophene-3-carboxylic acid
5-Methyl-thiophene-3-carboxylic acid
Formula: C₆H₆O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD01680229
CAS Number: 19156-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR