1-tert-Butyl-4-ethynylbenzene 97%

Supplier: Apollo Scientific
Warning

OR5953-25G OR5953-5G OR5953-1G
APOSOR5953-25GEA 132 EUR
APOSOR5953-25G APOSOR5953-5G APOSOR5953-1G
1-tert-Butyl-4-ethynylbenzene 97%
1-tert-Butyl-4-ethynylbenzene
Formula: C₁₂H₁₄
Boiling Pt: 70 °C/2mmHg
Density: 0,877 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00190197
CAS Number: 772-38-3
UN: 3082
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR