(2-Hydroxyphenyl)acetic acid 98%

Supplier: Apollo Scientific
Warning

OR5987-25G OR5987-100G OR5987-500G
APOSOR5987-25GEA 21.9 EUR
APOSOR5987-25G APOSOR5987-100G APOSOR5987-500G
(2-Hydroxyphenyl)acetic acid 98%
(2-Hydroxyphenyl)acetic acid
Formula: (C₆H₄OH)CH₂COOH
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
EINECS: 210-393-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR