5-Bromo-2-hydrazino-4-methylpyridine

Supplier: Apollo Scientific
Warning

OR60233-5G OR60233-1G
APOSOR60233-5GEA 546 EUR
APOSOR60233-5G APOSOR60233-1G
5-Bromo-2-hydrazino-4-methylpyridine
5-Bromo-2-hydrazino-4-methylpyridine
Formula: C₆H₈BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD09999921
CAS Number: 913839-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR