5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-1-methyl-2-oxoindoline, 5-Bromo-1-methyl-2-oxindole

OR6071-1G
APOSOR6071-1GEA 34.8 EUR
APOSOR6071-1G
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one 97%
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one
Formula: C₉H₈BrNO
MW: 226.08 g/mol
Melting Pt: 132…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00464061
CAS Number: 20870-90-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR