2-Amino-4,6-dimethylpyridine 97%

Supplier: Apollo Scientific
Warning

OR6121-1G OR6121-25G OR6121-5G
APOSOR6121-1GEA 21.9 EUR
APOSOR6121-1G APOSOR6121-25G APOSOR6121-5G
2-Amino-4,6-dimethylpyridine 97%
2-Amino-4,6-dimethylpyridine
Formula: C₇H₁₀N₂
Boiling Pt: 235 °C
Melting Pt: 63…64 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006322
CAS Number: 5407-87-4
EINECS: 226-470-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR