(Bromoethynyl)triisopropylsilane ≥97%

Supplier: Apollo Scientific

OR62151-1G OR62151-5G OR62151-10G
APOSOR62151-1GEA 43.5 EUR
APOSOR62151-1G APOSOR62151-5G APOSOR62151-10G
(Bromoethynyl)triisopropylsilane ≥97%
(Bromoethynyl)triisopropylsilane
Formula: C₁₁H₂₁BrSi
Density: 1,09
MDL Number: MFCD11036302
CAS Number: 111409-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR