4-(Furan-2-yl)pyridine-2-carboxylic acid

Supplier: Apollo Scientific
Warning

OR6253-1G OR6253-250MG OR6253-500MG
APOSOR6253-1GEA 478 EUR
APOSOR6253-1G APOSOR6253-250MG APOSOR6253-500MG
4-(Furan-2-yl)pyridine-2-carboxylic acid
4-(Furan-2-yl)pyridine-2-carboxylic acid
Formula: C₁₀H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD08056303
CAS Number: 914637-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR