Aminoacetaldehyde diethyl acetal

Supplier: Apollo Scientific
Danger

Synonyms: 2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal

OR6496-5G
APOSOR6496-5GEA 21.9 EUR
APOSOR6496-5G
Aminoacetaldehyde diethyl acetal
Aminoacetaldehyde diethyl acetal
Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
Boiling Pt: 161…163 °C
Melting Pt: –78 °C
Density: 0.916 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008136
CAS Number: 645-36-3
EINECS: 211-439-4
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR