6-Methoxy-1-tetralone 98%

Supplier: Apollo Scientific
Warning

OR6567-5G OR6567-1G OR6567-25G OR6567-100G
APOSOR6567-5GEA 0 EUR
APOSOR6567-5G APOSOR6567-1G APOSOR6567-25G APOSOR6567-100G
6-Methoxy-1-tetralone 98%
6-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
Boiling Pt: 171 °C/11mmHg
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR