2-Nitro-p-toluidine 98+%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Methyl-2-nitroaniline, 4-Amino-3-nitrotoluene, 2-Nitro-4-methylaniline

OR6639-100G OR6639-2.5KG OR6639-500G
APOSOR6639-100GEA 71.5 EUR
APOSOR6639-100G APOSOR6639-2.5KG APOSOR6639-500G
2-Nitro-p-toluidine 98+%
2-Nitro-p-toluidine
Formula: C₇H₈N₂O₂
Boiling Pt: 169 °C/12mmHg
Melting Pt: 115…118 °C
Density: 1,31 g/cm³ (25 °C)
Flash Pt: 157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007907
CAS Number: 89-62-3
EINECS: 201-924-9
UN: 2660
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR