2-Bromo-5-chlorobenzaldehyde

Supplier: Apollo Scientific
Warning

OR6701-100G OR6701-25G OR6701-500G
APOSOR6701-100GEA 94.4 EUR
APOSOR6701-100G APOSOR6701-25G APOSOR6701-500G
2-Bromo-5-chlorobenzaldehyde
2-Bromo-5-chlorobenzaldehyde
Formula: C₇H₄BrClO
Melting Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00462870
CAS Number: 174265-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR