(±)-β-Methylphenethylamine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Phenylpropylamine

OR6734-1G OR6734-25G OR6734-5G
APOSOR6734-1GEA 77 EUR
APOSOR6734-1G APOSOR6734-25G APOSOR6734-5G
(±)-β-Methylphenethylamine 98%
(±)-β-Methylphenethylamine
Formula: C₉H₁₃N
Boiling Pt: 82…84 °C/10mmHg
Density: 0,93 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008144
CAS Number: 582-22-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR