1-[2-(Morpholin-4-yl)ethyl]piperazine 98%

Supplier: Apollo Scientific
Danger

OR6912-1G OR6912-25G OR6912-5G
APOSOR6912-1GEA 21.9 EUR
APOSOR6912-1G APOSOR6912-25G APOSOR6912-5G
1-[2-(Morpholin-4-yl)ethyl]piperazine 98%
1-[2-(Morpholin-4-yl)ethyl]piperazine
Formula: C₁₀H₂₁N₃O
Boiling Pt: 106…107 °C/0,2mmHg
Storage Temperature: Ambient
MDL Number: MFCD00191217
CAS Number: 4892-89-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR