Diisopropyl(2-piperazin-1-ylethyl)amine 97%

Supplier: Apollo Scientific
Danger

OR6919-10G OR6919-2G OR6919-5G
APOSOR6919-10GEA 242 EUR
APOSOR6919-10G APOSOR6919-2G APOSOR6919-5G
Diisopropyl(2-piperazin-1-ylethyl)amine 97%
Diisopropyl(2-piperazin-1-ylethyl)amine
Formula: C₁₂H₂₇N₃
Boiling Pt: 64…66 °C/0,1mmHg
Storage Temperature: Ambient
MDL Number: MFCD01320902
CAS Number: 59955-93-0
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR