5-Nitro-2,4-xylidine

Supplier: Apollo Scientific
Warning

OR6966-1G OR6966-25G OR6966-5G
APOSOR6966-1GEA 21.9 EUR
APOSOR6966-1G APOSOR6966-25G APOSOR6966-5G
5-Nitro-2,4-xylidine
5-Nitro-2,4-xylidine
Formula: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00025199
CAS Number: 2124-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR