Methyl 3-cyano-5-nitrobenzoate

Supplier: Apollo Scientific
Warning

OR70185-5G
APOSOR70185-5GEA 97.3 EUR
APOSOR70185-5G
Methyl 3-cyano-5-nitrobenzoate
Methyl 3-cyano-5-nitrobenzoate
Formula: C₉H₆N₂O₄
MW: 206.155 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00458621
CAS Number: 99066-80-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR