4-Bromo-2-methoxy-1-methylsulphanylbenzene 95%

Supplier: Apollo Scientific

OR71005-1G OR71005-500MG
APOSOR71005-1GEA 161 EUR
APOSOR71005-1G APOSOR71005-500MG
4-Bromo-2-methoxy-1-methylsulphanylbenzene 95%
4-Bromo-2-methoxy-1-methylsulphanylbenzene
Formula: C₈H₉BrOS
Storage Temperature: Refrigerator
MDL Number: MFCD27944804
CAS Number: 144369-82-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR