1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%

Supplier: Apollo Scientific

OR7237-1G OR7237-5G
APOSOR7237-1GEA 238 EUR
APOSOR7237-1G APOSOR7237-5G
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester
Formula: C₁₄H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD06797198
CAS Number: 905966-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR