4'-Acetylbiphenyl-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR7314-1G OR7314-250MG OR7314-5G
APOSOR7314-1GEA 46.4 EUR
APOSOR7314-1G APOSOR7314-250MG APOSOR7314-5G
4'-Acetylbiphenyl-4-carboxylic acid
4'-Acetylbiphenyl-4-carboxylic acid
Formula: C₁₅H₁₂O₃
Melting Pt: 281…284 °C
Storage Temperature: Ambient
MDL Number: MFCD00230828
CAS Number: 114691-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR