[(1-Methylpiperidin-4-yl)methyl]amine dihydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: (1-Methylpiperidin-4-yl)methanamine dihydrochloride, 4-(Aminomethyl)-1-methylpiperidine dihydrochloride

OR7315-1G
APOSOR7315-1GEA 69.6 EUR
APOSOR7315-1G
[(1-Methylpiperidin-4-yl)methyl]amine dihydrochloride
[(1-Methylpiperidin-4-yl)methyl]amine dihydrochloride
Formula: C₇H₁₈Cl₂N₂
MW: 201.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035762
CAS Number: 1187582-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR