2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate 95+%

Supplier: Apollo Scientific
Warning

OR7624-1G OR7624-5G
APOSOR7624-1GEA 265 EUR
APOSOR7624-1G APOSOR7624-5G
2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate 95+%
2-Chloro-4-(methylsulphonyl)benzaldehyde hydrate
Formula: C₈H₇ClO₃S·H₂O
Melting Pt: 111…112 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08166750
CAS Number: 1171826-50-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR