1-[4-(Methylsulphonyl)-2-nitrophenyl]piperazine

Supplier: Apollo Scientific
Warning

OR7796-1G OR7796-250MG
APOSOR7796-1GEA 197 EUR
APOSOR7796-1G APOSOR7796-250MG
1-[4-(Methylsulphonyl)-2-nitrophenyl]piperazine
1-[4-(Methylsulphonyl)-2-nitrophenyl]piperazine
Formula: C₁₁H₁₅N₃O₄S
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD03407994
CAS Number: 610275-77-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR