3,5-Bis(methylsulphonyl)benzoic acid 95+%

Supplier: Apollo Scientific
Warning

OR7822-1G OR7822-5G OR7822-10G
APOSOR7822-1GEA 235 EUR
APOSOR7822-1G APOSOR7822-5G APOSOR7822-10G
3,5-Bis(methylsulphonyl)benzoic acid 95+%
3,5-Bis(methylsulphonyl)benzoic acid
Formula: C₉H₁₀O₆S₂
MW: 278.3 g/mol
Melting Pt: 283…285 °C
Storage Temperature: Ambient
MDL Number: MFCD04037979
CAS Number: 90536-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR