1H-Indene-3-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

OR7859-5G OR7859-10G
APOSOR7859-5GEA 219 EUR
APOSOR7859-5G APOSOR7859-10G
1H-Indene-3-carboxylic acid 95%
1H-Indene-3-carboxylic acid
Formula: C₁₀H₈O₂
Melting Pt: 161…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00086207
CAS Number: 14209-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR