[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine 95+%

Supplier: Apollo Scientific
Warning

OR7917-1G OR7917-5G OR7917-25G
APOSOR7917-1GEA 285 EUR
APOSOR7917-1G APOSOR7917-5G APOSOR7917-25G
[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine 95+%
[2-Chloro-5-methyl-4-(methylsulphonyl)phenyl]hydrazine
Formula: C₈H₁₁ClN₂O₂S
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD04037956
CAS Number: 849035-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR