5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-(2,6-Dimethylmorpholin-4-yl)-5-nitrobenzoic acid

OR8191-1G OR8191-250MG
APOSOR8191-1GEA 87 EUR
APOSOR8191-1G APOSOR8191-250MG
5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid
5-Nitro-2-(2,6-dimethylmorpholin-4-yl)benzoic acid
Formula: C₁₃H₁₆N₂O₅
MW: 280.28 g/mol
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00052231
CAS Number: 175136-71-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR