3-Bromo-2,6-dihydroxy-5-nitrobenzonitrile

Supplier: Apollo Scientific
Warning

OR8226-1G OR8226-25G OR8226-5G
APOSOR8226-1GEA 21.9 EUR
APOSOR8226-1G APOSOR8226-25G APOSOR8226-5G
3-Bromo-2,6-dihydroxy-5-nitrobenzonitrile
3-Bromo-2,6-dihydroxy-5-nitrobenzonitrile
Formula: C₇H₄BrNO
Melting Pt: 160 °C
Storage Temperature: Ambient
CAS Number: 288067-35-6
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR