8-Hydroxyquinoline-N-Oxide

Supplier: Apollo Scientific

OR8255-100G
APOSOR8255-100GEA 148 EUR
APOSOR8255-100G
8-Hydroxyquinoline-N-Oxide
8-Hydroxyquinoline-N-Oxide
Formula: C₉H₇NO₂
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00006739
CAS Number: 1127-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR