Risocaine (propyl-4-aminobenzoate)

Supplier: Apollo Scientific
Warning

OR8358-50G
APOSOR8358-50GEA 109 EUR
APOSOR8358-50G
Risocaine (propyl-4-aminobenzoate)
Risocaine (Propyl 4-aminobenzoate)
Formula: C₁₀H₁₃NO₂
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00017111
CAS Number: 94-12-2
EINECS: 202-306-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR