Trichloroacetic acid (TCA) 99%

Supplier: Apollo Scientific
Danger

Synonyms: TCA

OR8372-1KG OR8372-250G OR8372-5KG
APOSOR8372-1KGEA 139 EUR
APOSOR8372-1KG APOSOR8372-250G APOSOR8372-5KG
Trichloroacetic acid (TCA) 99%
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196 °C
Melting Pt: 52…58 °C
Density: 1.62g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 1839
ADR: 8 II
Merck Index: 13,09700

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR