α-Bromo-4-iodotoulene 97%

Supplier: Apollo Scientific
Danger

OR8750-100G OR8750-25G OR8750-5G
APOSOR8750-100GEA 244 EUR
APOSOR8750-100G APOSOR8750-25G APOSOR8750-5G
α-Bromo-4-iodotoulene 97%
α-Bromo-4-iodotoulene
Formula: C₇H₆BrI
Boiling Pt: 140 °C/15mmHg
Melting Pt: 78…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR