4-Acetyl-2-chloropyridine

Supplier: Apollo Scientific
Warning

Synonyms: 1-(2-chloropyridin-4-yl)ethan-1-one, 1-(2-chloropyridine-4-yl)ethanone

OR8761-1G
APOSOR8761-1GEA 21.9 EUR
APOSOR8761-1G
4-Acetyl-2-chloropyridine
4-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.58 g/mol
Boiling Pt: 96 °C/1.99mmHg
Melting Pt: 35…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07699406
CAS Number: 23794-15-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR