2-Methylpiperidine 99%

Supplier: Apollo Scientific
Danger

OR8797-100ML
APOSOR8797-100MLEA 29 EUR
APOSOR8797-100ML
2-Methylpiperidine 99%
2-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 117…119 °C
Melting Pt: –4 °C
Density: 0,844 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00005982
CAS Number: 109-05-7
UN: 2733
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR