4-Nitroanthranilic acid 98+%

Supplier: Apollo Scientific
Warning

OR8876-100G OR8876-25G OR8876-500G
APOSOR8876-100GEA 55.1 EUR
APOSOR8876-100G APOSOR8876-25G APOSOR8876-500G
4-Nitroanthranilic acid 98+%
4-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
Melting Pt: 257 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007262
CAS Number: 619-17-0
EINECS: 210-583-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR