Syringic acid 97%

Supplier: Apollo Scientific
Warning

OR8937-5G
APOSOR8937-5GEA 21.9 EUR
APOSOR8937-5G
Syringic acid 97%
Syringic acid
Formula: C₉H₁₀O₅
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR