Aniline hydrochloride

Supplier: Apollo Scientific
Danger

Synonyms: Phenylamine HCl, Aminobenzene hydrochloride, Anilinium chloride, Aminobenzene HCl, Phenylamine hydrochloride

OR8982-100G OR8982-2.5KG OR8982-500G
APOSOR8982-100GEA 90.2 EUR
APOSOR8982-100G APOSOR8982-2.5KG APOSOR8982-500G
Aniline hydrochloride
Aniline hydrochloride
Formula: C₆H₈ClN
MW: 129.59 g/mol
Boiling Pt: 245 °C
Melting Pt: 196…200 °C
Density: 1.221 g/cm³ (25 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00012958
CAS Number: 142-04-1
UN: 1548
ADR: 6.1 III
Merck Index: 13,00616

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR