1H-Indene-2-carboxylic acid 96%

Supplier: Apollo Scientific

OR900045-1G OR900045-250MG
APOSOR900045-1GEA 632 EUR
APOSOR900045-1G APOSOR900045-250MG
1H-Indene-2-carboxylic acid 96%
1H-Indene-2-carboxylic acid
Formula: C₁₀H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00039586
CAS Number: 41712-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR