N-tert-Butyl-4-hydroxybenzamide 98%

Supplier: Apollo Scientific

OR900667-1G
APOSOR900667-1GEA 270 EUR
APOSOR900667-1G
N-tert-Butyl-4-hydroxybenzamide 98%
N-tert-Butyl-4-hydroxybenzamide
Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD11130632
CAS Number: 250656-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR