1-Benzylpyrrolidin-3-ylmethanol 97%

Supplier: Apollo Scientific

OR901559-25G OR901559-5G
APOSOR901559-25GEA 401 EUR
APOSOR901559-25G APOSOR901559-5G
1-Benzylpyrrolidin-3-ylmethanol 97%
1-Benzylpyrrolidin-3-ylmethanol
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD04038527
CAS Number: 5731-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR