2-Methoxy-6-p-tolylpyridine 97%

Supplier: Apollo Scientific

OR901891-1G
APOSOR901891-1GEA 364 EUR
APOSOR901891-1G
2-Methoxy-6-p-tolylpyridine 97%
2-Methoxy-6-p-tolylpyridine
Formula: C₁₃H₁₃NO
MW: 199,248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18914538
CAS Number: 1039775-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR