3-Methoxy-2-nitrobenzaldehyde 95%

Supplier: Apollo Scientific

OR902223-5G
APOSOR902223-5GEA 217 EUR
APOSOR902223-5G
3-Methoxy-2-nitrobenzaldehyde 95%
3-Methoxy-2-nitrobenzaldehyde
Formula: C₈H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00007135
CAS Number: 53055-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR