L-(+)-Methyl-tert-leucinate hydrochloride 97%

Supplier: Apollo Scientific

OR903118-100G OR903118-25G OR903118-5G
APOSOR903118-100GEA 405 EUR
APOSOR903118-100G APOSOR903118-25G APOSOR903118-5G
L-(+)-Methyl-tert-leucinate hydrochloride 97%
L(+)-Methyl tert-leucinate hydrochloride
Formula: C₇H₁₅NO₂·HCl
Melting Pt: 183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040604
CAS Number: 63038-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR