2-Methoxy-5-(2-pyrrolidinyl)pyridine 95%

Supplier: Apollo Scientific

OR904134-1G OR904134-250MG
APOSOR904134-1GEA 533 EUR
APOSOR904134-1G APOSOR904134-250MG
2-Methoxy-5-(2-pyrrolidinyl)pyridine 95%
2-Methoxy-5-(2-pyrrolidinyl)pyridine
Formula: C₁₀H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD05189295
CAS Number: 185510-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR