Thiophene-2-carboxaldoxime 95%

Supplier: Apollo Scientific

OR904201-25G
APOSOR904201-25GEA 84.1 EUR
APOSOR904201-25G
Thiophene-2-carboxaldoxime 95%
Thiophene-2-carboxaldoxime
Formula: C₅H₅NOS
Storage Temperature: Ambient
MDL Number: MFCD00022509
CAS Number: 29683-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR