5-Bromo-1-pentanol 97%

Supplier: Apollo Scientific

OR907161-100G OR907161-25G OR907161-5G
APOSOR907161-100GEA 151 EUR
APOSOR907161-100G APOSOR907161-25G APOSOR907161-5G
5-Bromo-1-pentanol 97%
5-Bromo-1-pentanol
Formula: C₅H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD00061101
CAS Number: 34626-51-2
UN: 1993
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR