1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one 95%

Supplier: Apollo Scientific

OR908611-250MG
APOSOR908611-250MGEA 957 EUR
APOSOR908611-250MG
1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one 95%
1-Cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1h)-one
Formula: C₁₄H₂₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16996103
CAS Number: 1596367-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR