Triphenylphosphine 99+%

Supplier: Apollo Scientific
Warning

OR9088-1KG OR9088-250G OR9088-5KG
APOSOR9088-1KGEA 78.3 EUR
APOSOR9088-1KG APOSOR9088-250G APOSOR9088-5KG
Triphenylphosphine 99+%
Triphenylphosphine
Formula: C₁₈H₁₅P
Boiling Pt: 360 °C
Melting Pt: 78…82 °C
Density: 1,2 g/cm³ (25 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00003043
CAS Number: 603-35-0
EINECS: 210-036-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR