Cyclopropylethynylboronic acid pinacol ester 95%

Supplier: Apollo Scientific

Synonyms: 2-(Cyclopropylethynyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR908926-1G
APOSOR908926-1GEA 256 EUR
APOSOR908926-1G
Cyclopropylethynylboronic acid pinacol ester 95%
Cyclopropylethynylboronic acid pinacol ester
Formula: C₁₁H₁₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD13182125
CAS Number: 1432491-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR