1-Methyl-1H-indazole 98%

Supplier: Apollo Scientific
Warning

OR910488-1G
APOSOR910488-1GEA 75.4 EUR
APOSOR910488-1G
1-Methyl-1H-indazole 98%
1-Methyl-1H-indazole
Formula: C₈H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00661658
CAS Number: 13436-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR